Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Clear Silicone Rubber, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
24234-C
102092-302EA 422.6 USD InStock
102092-302
Clear Silicone Rubber, Electron Microscopy Sciences
Specimen Mounting Supplies
This silicone cures at room temperature with no shrinkage and offers a unique water clear see through rubber with excellent tear strength, chemical and heat resistance.

Curing: allow the mold to cure overnight (at least 16 hours) at room temperature (77°F/25°C) before demolding. Do not cure rubber where temperature is less than 65°F/18°C. Post Curing the mold will aid in quickly attaining maximum physical and performance properties. After curing at room temperature, expose the rubber to 80°C for 2 hours and 100°C for 2 hours. Allow mold to cool to room temperature before using.

Technical Overview
Hardness Shore A 6: 40
Color: Translucent
Pot Life: 60 min.
Demold Time: 16 hours
Specific Volume: 25.8
Specific Gravity: 1.07
Mixed Viscosity: 35,000cps
Die B Tear Strength: 120 pli
Tensile Strength: 800 psi
Shrinkage: Negligible
Mixing Ratio Wt. Or Vol.: (100:10) pbw

Ordering information: Clear silicone comes as a kit of 1 gallon (9 lb) silicone A and 1 pt (.9lb) curing agent B.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty