Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Anti-ESR Mouse Monoclonal Antibody [clone: 6F11]

Supplier: Diagnostic BioSystems

PDM048 MOB121 MOB121 PDM048
99990-890EA 719.16 USD InStock
99990-890 CA99990-890 99990-316 CA99990-316
Anti-ESR Mouse Monoclonal Antibody [clone: 6F11]
Antibodies
This antibody is specific to estrogen receptor, which is associated with superior prognosis and a better response to anti-estrogen therapy. The nuclei of the estrogen receptor positive cells stain very strongly with this antibody, without any staining in the cytoplasm.

Type:
Antigen: ESR
Clonality: Monoclonal
Clone: 6F11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty