Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Anti-ESR Rabbit Monoclonal Antibody [clone: SP1]

Supplier: Diagnostic BioSystems

RMAB001 RMPD001 RMPD001 RMAB001
99991-128EA 2347.49 USD InStock
CA99991-128 99991-128 CA99991-164 99991-164
Anti-ESR Rabbit Monoclonal Antibody [clone: SP1]
Antibodies
This antibody recognizes a protein of 67kDa, which is identified as estrogen receptor (ER). ER content of breast cancer tissue is an important parameter in the prediction of prognosis and response to endocrine therapy. This antibody strongly stains the nucleus of epithelial cells in breast carcinomas. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Type:
Antigen: ESR
Clonality: Monoclonal
Clone: SP1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty