Would you like to log in again?
Iron Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic BioSystems
KT021 KT021
95027-200EA 462.43 USD InStock
CA95027-200 95027-200
Iron Stain Kit, Diagnostic Biosystems
Stains Histology Stains
This kit stains ferric iron in paraffin tissue sections
The iron deposits stain blue, while the nuclei and other tissue stain red and pink respectively.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty