Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Solvent Blue 38 kit stain for histology

Supplier: Diagnostic Biosystems

Synonyms: Direct Blue 86Luxol® Fast Blue MBSNC.I. 74180

KT022 KT022
95027-202EA 693.64 USD InStock
95027-202 CA95027-202
Solvent Blue 38 kit stain for histology
Solvent Blue 38
This kit stains myelin sheath and Nissl substance in paraffin tissue sections

The myelin fibers stain blue to greenish blue, while the cells stain pink to violet.


Formula: C₃₂H₁₂CuN₈Na₂O₆S₂
MW: 778.15 g/mol
MDL Number: MFCD00071424
CAS Number: 1328-51-4
Order Now


 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty