Would you like to log in again?
HyperSep™ C8 SPE Cartridges
Supplier: Thermo Scientific
HyperSep™
60108-394 60108-393 60108-391 60108-392 60108-427 60108-309 60108-706 60108-704
10047-252EA 285.73 USD InStock
10047-252 10047-258 10047-346 10047-228 10047-254 10047-298 10047-342 10047-256
HyperSep™ C8 SPE Cartridges
Chromatography Columns
HyperSep™ C8 cartridges, less retentive alternative to C18 for polar and nonpolar compounds.
  • Well suited for method requiring less retention than C18
  • For moderately polar analytes
  • Potential for polar interactions greater than with C18
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Packaging: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty