Would you like to log in again?
HyperSep™ Florisil SPE Cartridges
Supplier: Thermo Scientific
HyperSep™
60108-431 60108-402 60108-737 60108-735 60108-500 60108-736 60108-403 60108-405 60108-404
10047-264EA 480.32 USD InStock
10047-264 10048-108 10047-308 10047-270 10048-110 10048-106 10047-266 10047-268 10047-300
HyperSep™ Florisil SPE Cartridges
Chromatography Columns
Ideal for the isolation of polar compounds from non-polar matrices.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty