Would you like to log in again?
HyperSep™ Diol SPE Cartridges
Supplier: Thermo Scientific
HyperSep™
60108-571 60108-572 60108-574 60108-576 60108-757 60108-573 60108-755 60108-756 60108-575
10047-330EA 293.78 USD InStock
10047-330 10048-142 10047-328 10048-146 10047-324 10048-144 10047-326 10047-332 10047-322
HyperSep™ Diol SPE Cartridges
Chromatography Columns
A short alkyl chains with polar functional groups are ideal for the isolation of polar compounds.
  • Normal phase extraction of polar compounds
  • Purification of polar compounds
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Packaging: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty