Would you like to log in again?
HyperSep™ SLE Cartridges, Thermo Scientific
Supplier: Thermo Scientific
HyperSep™
60109-20000609 60109-500-3-7 60109-4000-259 60109-200-3-9 60109-2000-129 60109-200-3-7 60109-1000-6-7 60109-4000-257 60109-500-6-9 60109-500-3-9 60109-20000607 60109-500-6-7 60109-2000-127 60109-1000-6-9
10048-602EA 257.22 USD InStock
10048-602 10047-930 10047-940 10047-942 10047-950 10047-948 10047-956 10047-944 10048-592 10047-958 10048-590 10047-946 10048-596 10048-594
HyperSep™ SLE Cartridges, Thermo Scientific
Chromatography Columns
Solid supported liquid/liquid extraction (SLE) is a fast effective sample preparation technique that provides considerable benefits over liquid-liquid extraction (LLE) protcols for removal of phospholipids, from biological samples. HyperSep™ SLE deals with sample preparation of biological matricies via a simple, efficient and an effective protocol, the focus being the removal of interfering phospholipids which cause a number of issues for end point analysis.
  • Simple, easy to use protocol
  • Removal of phospholipids
  • Greater reproducibility and recoveries compared to LLE
SLE offers a number of advantages over existing sample preparation techniques such as Liquid/Liquid Extraction (LLE) and protein precipitation techniques for dealing with biological matricies. In comparison to LLE, SLE offers greater reproducibility and recoveries, lower solvent requirements and prevents emulsification which is often associated with the LLE technique. It also lends itself to full automation which is not possible via LLE. SLE provides improved cleanliness of sample extract and associated increase in sensitivity compared to protein precipitation techniques which are unable to remove phospholipids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty