Would you like to log in again?
Chloride Reagent Set, Drop Count Titration, Hach
Supplier: Hach
2437600
89231-074EA 99.74 USD InStock
89231-074
Chloride Reagent Set, Drop Count Titration, Hach
Environmental Test Kits
For determination of chloride by argentometric drop-count titration
Replacement reagent set for 8-P Test Kit. Set includes Chloride 2 Indicator Powder Pillows and Silver Nitrate Titrant (0.0493 N titrant). Approx. 100 tests.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty