Would you like to log in again?
Chlorine (Total) Reagent Set, HR Digital Titrator, Hach
Supplier: Hach
2444800 2444800
89231-392EA 240.79 USD InStock
CA11024-844 89231-392
Chlorine (Total) Reagent Set, HR Digital Titrator, Hach
Environmental Test Kits
For determination of HR Total Chlorine by Digital Titration with Sodium Thiosulfate. Range: 2000 to 70,000 mg/L Cl2. Method: Iodometric Set includes Dissolved Oxygen 3 Powder Pillows, Potassium Iodide Powder Pillows, Starch Indicator Solution and Sodium Thiosulfate Digital Titratior Cartridges (2) 2.00 N. Approximately 100 tests.
  • Accurate Results; when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical
Ordering information: Hazardous Items; items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty