Would you like to log in again?
E-A-R™ Classic Soft™ Earplugs, 3M™
Supplier: 3M
E-A-R™
7000127652
89006-758BX 115.88 USD InStock
89006-758
E-A-R™ Classic Soft™ Earplugs, 3M™
Earplugs Disposable Earplugs
These earplugs feature a unique PU-foam combination which warms to the body temperature and becomes softer, thus allowing the earplug to mould to the individuals ear canal shape.
  • Efficient seal due to the moulding of the earplug to the ear canal
  • Textured surface provides greater friction
  • Improves grip and helps maintain an effective seal
Resistant to moisture for hot and humid working environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty