Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1000 Watt Type 3 Voltage Converter, Step Up and Step Down Voltage Transformer

Supplier: INTERNATIONAL VIDEO & ELECTRONICS
DS-1000
470061-810EA 172.6 USD InStock
470061-810
1000 Watt Type 3 Voltage Converter, Step Up and Step Down Voltage Transformer
Power Supplies
The International DS-1000 Diamond Series 1000 Watt Type 3 Voltage converters offer best in class performance in voltage transformers. They are thoroughly tested to be free of defects in workmanship and materials and hand soldering insures to meet U.S Military specifications.

  • USA grounded male plug
  • Extra adapter included to use with 220 V outlets
  • Converts 110/120 V up to 220/240 V and vice versa
  • Works anywhere in the world
  • Heavy duty for continuous use
  • Extra durable lifetime coil is included
  • Recommended for high value and/or sensitive items

Weight: 15 lbs. (6.8 kg)
Dimensions: 7.25x8.25x5.75" (18.4x21x14.6 cm)

Ideal when using a ¼" drill, a clothes dryer or an LCD TV or when moving or visiting other countries. BEST Diamond series 5 year warranty.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty