Would you like to log in again?
SP Bel-Art Glove Box Sleeved Gloves, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
H50025-0410 H50025-0409
47735-532PK 652.08 USD InStock
47735-532 47735-530
SP Bel-Art Glove Box Sleeved Gloves, Bel-Art Products, a part of SP
Gloves
One-piece, seamless, smooth neoprene protection against liquid, gas, or solid substance penetration.
  • One-piece design
  • Seamless
  • Smooth neoprene protection
  • Must be used with clamping rings
Suitable for use with glove boxes having 8" glove ports including Clear View or sidENTRY™ Glove Boxes. Requires clamping rings (47748-192) to hold gloves in place.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty