Would you like to log in again?
VWR® Lister Bandage Scissors
Supplier: Avantor
11648-341
89259-978EA 37.34 USD InStock
89259-978
VWR® Lister Bandage Scissors
Scissors
VWR® Lister Bandage Scissors are stainless steel with one blade with a rounded tip and one blade with a blunt, flattened, probe tip.
  • Scissors are angular
Lister bandage scissors are available in a range of lengths and order sizes, from individual to bulk.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty