Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDKN1B Mouse Monoclonal Antibody [clone: DCS-72.F6]

Supplier: Diagnostic BioSystems

MOB281-05 MOB281
99990-532EA 878.13 USD InStock
99990-532 95024-738
Anti-CDKN1B Mouse Monoclonal Antibody [clone: DCS-72.F6]
Antibodies
This antibody is specific to a protein of 27 kDa known as p27Kip1 a cell cycle regulatory mitotic inhibitor. This antibody recognizes an epitope between amino acids 83-204 of p27. This antibody does not show any cross-reactivity with other related mitotic inhibitors.

Type:
Antigen: CDKN1B
Clonality: Monoclonal
Clone: DCS-72.F6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty