Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Prohibitin Mouse Monoclonal Antibody [clone: II-14-10]

Supplier: Diagnostic BioSystems

MOB323 PDM152 PDM152 MOB323-05
95029-778EA 888.84 USD InStock
95029-778 95024-796 CA95029-778 99990-604
Anti-Prohibitin Mouse Monoclonal Antibody [clone: II-14-10]
Antibodies
This antibody is specific to a 30 kDa protein known as prohibitin. Prohibitin is an evolutionarily conserved protein located in the inner membrane of mitochondria. Prohibitin shows antiproliferative activity and has been proposed to play a role in normal cell cycle regulation, replicative senescence, cellular immortalization and tumor suppression.

Type:
Antigen: Prohibitin
Clonality: Monoclonal
Clone: II-14-10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty