Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Melanoma Mouse Monoclonal Antibody [clone: HMB45]

Supplier: Diagnostic BioSystems

MOB079 MOB079-05 PDM011
99990-262EA 691.83 USD InStock
99990-262 95024-668 99990-834
Anti-Melanoma Mouse Monoclonal Antibody [clone: HMB45]
Antibodies
This antibody reacts with a neuraminidase sensitive oligosaccharide side chain of a glycoconjugate present in immature melanosomes. It reacts with junctional and blue nevus cells. Non-melanocytic cells are negative.

Type:
Antigen: Melanoma
Clonality: Monoclonal
Clone: HMB45
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty