Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Melanoma Mouse Monoclonal Antibody [clone: PNL2]

Supplier: Diagnostic BioSystems

MOB421-05 MOB421
95024-670EA 700.53 USD InStock
95024-670 99990-778
Anti-Melanoma Mouse Monoclonal Antibody [clone: PNL2]
Antibodies
This antibody labels melanocytes and is a useful tool for the identification of melanomas and clear cell sarcomas. Anti-Melanoma, PNL2, seems to be a melanocyte antibody of autoimmune origin as it does not react with the non-melanocytic peptide antigen used for immunization.

Type:
Antigen: Melanoma
Clonality: Monoclonal
Clone: PNL2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty