Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RPA/p34 Mouse Monoclonal Antibody [clone: 9H8]

Supplier: Diagnostic BioSystems

MOB329 MOB329-05
95024-816EA 557.69 USD InStock
95024-816 99990-616
Anti-RPA/p34 Mouse Monoclonal Antibody [clone: 9H8]
Antibodies
This antibody reacts with a 32-34 kDa protein known as replication protein A (RPA). RPA is involved in DNA replication, repair and recombination. Human RPA is a stable heterotrimer of 70 kDa, 32-34 kDa and 11-14 kDa subunits (RPA70, RPA32 and RPA14 respectively). RPA is required for the SV40 large tumor antigen-catalyzed unwinding for SV40 DNA and stimulates DNA polymerase α and δ.

Type:
Antigen: RPA/p34
Clonality: Monoclonal
Clone: 9H8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty