Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S100 Mouse Monoclonal Antibody [clone: SH-B1]

Supplier: Diagnostic BioSystems

PDM088 MOB111-05 MOB111
95024-818EA 368.89 USD InStock
95024-818 99990-306 99990-958
Anti-S100 Mouse Monoclonal Antibody [clone: SH-B1]
Antibodies
This antibody is specific against an epitope located on the β chain (i.e. in S-100a and S-100b) but not on the α chain of S-100 (i.e. in S-100a and S-100ao). This antibody can be used to localize S-100a and S-100b in various tissue sections.

Type:
Antigen: S100
Clonality: Monoclonal
Clone: SH-B1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty