Would you like to log in again?
TraceGOLD TG-XLBMS GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26079-1540 26079-1300 26079-1550 26079-2230 26079-1420 26079-1430 26079-5780
10055-526EA 746.83 USD InStock
10055-526 10055-536 10055-538 10055-532 10055-554 10055-534 10055-530
TraceGOLD TG-XLBMS GC Columns, Thermo Scientific
Chromatography Columns
General purpose columns exhibing extremely low bleed.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty