Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
TraceGOLD TG-17MS GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26089-1420 26089-1440 26089-2970 26089-2980 26089-2230 26089-1430 26089-1300 26089-3360 26089-2960 26089-2240
10055-866EA 1178.53 USD InStock
10055-866 10055-852 10055-840 10055-862 10055-832 10055-842 10055-864 10055-870 10055-850 10055-838
TraceGOLD TG-17MS GC Columns, Thermo Scientific
Chromatography Columns
Particular suited to GCMS applications that require more polarity than a 5% Phenyl phase.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty