Would you like to log in again?
TraceGOLD TG-624 GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26085-3410 26085-4950 26085-4080 26085-3960 26085-3320 26085-4090 26085-4900 26085-3330 26085-4960 26085-3390
10056-336EA 1119.08 USD InStock
10056-336 10056-346 10056-344 10056-338 10056-348 10056-350 10056-354 10056-342 10056-352 10056-340
TraceGOLD TG-624 GC Columns, Thermo Scientific
Chromatography Columns
Offers 90+% resolution of the first six gases in EPA Method 8260 and EPA Method 524.2 for volatile organics analysis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty