Would you like to log in again?
TraceGOLD TG-WaxMS A GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26087-2860 26087-1420 26087-2240 26087-1310 26087-1430 26087-2980 26087-2970 26087-1300 26087-2230 26087-1540 26087-1440
10055-702EA 767.97 USD InStock
10055-702 10055-704 10055-676 10055-688 10055-672 10055-674 10055-690 10055-706 10056-386 10055-670 10056-384
TraceGOLD TG-WaxMS A GC Columns, Thermo Scientific
Chromatography Columns
Acidic functionality in the polymer structure allows analysis of acidic compounds without derivatization.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty