Would you like to log in again?
TraceGOLD TG-WaxMS B GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26086-2980 26086-2230 26086-1420 26086-1430
10056-360EA 1143.15 USD InStock
10056-360 10056-376 10056-364 10056-362
TraceGOLD TG-WaxMS B GC Columns, Thermo Scientific
Chromatography Columns
Base deactivation allows analysis of basic analytes with derivatization or column priming.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty