Would you like to log in again?
TracePLOT TG-BOND Alumina GC Columns KCI Deactivation, Thermo Scientific
Supplier: Thermo Scientific
TracePLOT
26002-6110 26002-6050 26002-6020 26002-6080
10056-016EA 2373.74 USD InStock
10056-016 10056-014 10056-012 10056-010
TracePLOT TG-BOND Alumina GC Columns KCI Deactivation, Thermo Scientific
Chromatography Columns
Optimized for linear and quantitative analysis of polar unsaturated hydrocarbons.
  • Strong bonding to prevent particle generation
  • Columns to which water has adsorbed may be regenerated to restore full efficiency and selectivity
Each column has been tested to ensure proper film thickness (1,3-butadiene), selectivity (propadiene and methyl acetylene), resolution (trans-2-butene and 1-butene) and coating efficiency (1,3-butadiene).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty