Would you like to log in again?
TRACE TR-1MS GC Columns, Thermo Scientific
Supplier: Thermo Scientific
TRACE
260B309P 260B142P 260B047P 260B154P 260B155P 260B143P
10056-962EA 1873.36 USD InStock
10056-962 10056-960 10056-964 10056-958 10056-968 10056-970
TRACE TR-1MS GC Columns, Thermo Scientific
Chromatography Columns
Extremely low-bleed non-polar columns suitable for GC/MS applications.
  • Non-polar phase, 100% dimethyl polysiloxane
  • High operating temperature
  • Inert phase suited for environmental analyzes
Ideal for chlorinated and nitroaromatic compounds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty