Would you like to log in again?
TRACE TR-FAME GC Columns, Thermo Scientific
Supplier: Thermo Scientific
TRACE
260M155P 260M149P 260M154P 260M143P 260M147P 260M096P 260M142P 260M166L 260M141P
10057-256EA 1872.99 USD InStock
10057-256 10057-248 10057-250 10057-260 10057-252 10057-262 10057-240 10057-254 10057-242
TRACE TR-FAME GC Columns, Thermo Scientific
Chromatography Columns
TRACE™ TR-FAME GC columns are ideal for separation of cis or trans isomers and fatty acid methyl esters (FAMEs) with various degrees of unsaturation.
  • Low bleed for MS, even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance
  • Polar phase, 70% cyanopropyl polysilphenylene-siloxane
  • Higher operating temperature than other columns
These columns are also suitable for any application that requires a high-polarity column for use with MS detection.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty