Would you like to log in again?
Accucore™ XL C8 4 μm HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
Accucore™
74204-104630 74204-154630 74204-153030 74204-053030 74204-254630
10037-646EA 1214.01 USD InStock
10037-646 10038-654 10037-548 10037-650 10037-644
Accucore™ XL C8 4 μm HPLC Columns, Thermo Scientific
Chromatography Columns
Accucore™ XL C8 4 μm HPLC Columns offer lower hydrophobic retention than columns packed with longer alkyl chain length material, such as C18, and are therefore recommended for analytes with moderate hydrophobicity.
  • 4 μm Solid Core Particles for All Users
  • Same System, Same Method, Better Results
  • Robust, Fast, and Easy To Use
  • Similar Selectivity to C18, with Lower Retention
  • Recommended for Analytes with Moderate Hydrophobicity
Based on Core Enhanced Technology™ using 4 μm solid core particles, Accucore™ XL HPLC columns allow users of conventional HPLC methods to enjoy performance far beyond that of columns packed with 5, 4, or even 3 μm fully porous particles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty