Would you like to log in again?
Tygon S3™ B-44-4X I.B. Pressure Tubing, Saint-Gobain Performance Plastics
Supplier: Saint-Gobain
Tygon S3™
ADA00053 ADA00029 ADA05079 ADA06074 ADA00064 ADAUN029 ADA05074 ADA00018 ADAUN064 ADA00038 ADA05086 ADA00074 ADA00070 ADA00046
89404-252PK 1258.79 USD InStock
89404-252 89404-262 89404-254 89404-264 89404-242 89404-256 89404-244 89404-266 89404-258 89404-246 89404-270 89404-250 89404-260 89404-248
Tygon S3™ B-44-4X I.B. Pressure Tubing, Saint-Gobain Performance Plastics
Tubing Transfer Tubing
These tubings are smooth, with non-porous bore that resists particle entrapment.
  • Handles four times the pressure of non-reinforced tubing
  • Clear wall enables visualization of fluid flow
  • Connects easily to ReSeal® sanitary fittings
Certifications: Meets FDA, 3-A and NSF criteria
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty