Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium chloride 50 mM in aqueous solution

Supplier: MERIDIAN LIFE SCICNE, INC BE

BIO-37026
490005-950EA 83.9 USD InStock
490005-950
Magnesium chloride 50 mM in aqueous solution
Magnesium chloride
Composition, MgCl2 50 mM in nuclease free water. The 50 mM MgCl2 Solution should be stored at -20°C.


Formula: MgCl₂
MW: 95.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty