Would you like to log in again?
MBP SoftFit-L Tips, Thermo Scientific
Supplier: Thermo Scientific
3722-05
490008-506CS 927.69 USD InStock
490008-506
MBP SoftFit-L Tips, Thermo Scientific
Pipette Tips
ART Barrier SoftFit-L pipette tips seal when exposed to potential contaminants.
  • Available in MicroPoint to reduce surface area
  • Decrease sample retention for critical samples
  • Certified free from RNase, DNase and pyrogen
Designed for use with the Rainin LTS Litetouch pipettor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty