Would you like to log in again?
GeneMate Wide and Ultra-Wide Format Gel Systems, Galileo Bioscience
Supplier: Galileo Bioscience
81-2325 81-2340BEP 80-1410 81-2314
490001-342EA 1335.01 USD InStock
490001-342 490001-500 490001-504 490001-508
GeneMate Wide and Ultra-Wide Format Gel Systems, Galileo Bioscience
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Wide Horizontal Gel System:
  • Maxi-Wide Horizontal Gel System:
  • 500-sample capacity
  • 23 x 25 cm
  • Maxi-Plus Horizontal Gel System:
  • 600-sample capacity
  • 23 x 40 cm
Ultra-Wide Horizontal Gel System:
Large sample capacity
Rugged trouble-free construction
Multiple configurations for high throuhgput
ExpressCast end-gated trays for tape-free casting
Wide range of combs and accessories
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty