Would you like to log in again?
National Scientific Supply Filtered Pipette Tips
Supplier: National Scientific
AN0101-MRS
490009-594PK 727.69 USD InStock
490009-594
National Scientific Supply Filtered Pipette Tips
Pipette Tips
Non-wettable zero-fluid-retention pipette tips with beveled-ends ensure unsurpassed pipetting accuracy and reproducibility, allowing micro-volume dispensing without the need to touch-off.
  • Effectively eliminates unwanted signals induced by residual RNA/DNA transferred in trace amounts by aerosol cross-contamination
  • Also acts as a safety-barrier against accidental over-pipetting of liquid without risk of sample lock-up
  • Debris-free, non-reactive barriers are manufactured without surfactants, or other extractable components, to precise air-flow standards
Barriers block aerosols by coalescing radio-active, chemical, or biological contaminants before they can come in contact with pipettor shafts. Tips are specifically designed for use with Pipetman, Eppendorf, Finnpipette, and other research-grade pipettors.
Applications include any discrete sampling procedure requiring an aerosol-free system for transferring volatile or hazardous samples. Sterile.
Pipetman is a registered trademark of Gilson Medical Electronics (France) S.A. Eppendorf is a registered trademark of Eppendorf-Netheler-Hinz GmbH. Finnpipette is a registered trademark of Labsystems, Inc.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty