Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

IPTG (isopropyl-β-D-thiogalactopyranoside), Bioline

Supplier: MERIDIAN LIFE SCICNE, INC BE
Synonyms: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol

BIO-37036
490005-954EA 493.08 USD InStock
490005-954
IPTG (isopropyl-β-D-thiogalactopyranoside), Bioline
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Isopropyl-D-thiogalactopyranoside (IPTG) is a chemical analogue of galactose, which cannot be hydrolyzed by the enzyme --Galactosidase. Hence, it induces the E. coli lac operon activity by binding and inhibiting the lac repressor without being degraded.

  • Blue/White Color Screening
  • Induction of Lac Operon for Protein Expression
  • Genes Controlled by the Lac to Tac Promoter/Operator Sequences are Expressed to High Levels in the Presence of IPTG

Genes controlled by the lac or tac promotor/operator sequences are expressed to high levels in the presence of IPTG. Features include induces E. coli lac operon activity, >99.6% by HPLC, and available as powder and stabilized stock solution. IPTG powder can be stored for 12 months at -20°C. Shipped on dry ice or blue ice.


Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 109.5…110.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty