Would you like to log in again?
AluminaSeal Seals, Diversified Biotech
Supplier: Diversified Biotech
ALUM-1000
490006-674EA 77.04 USD InStock
490006-674
AluminaSeal Seals, Diversified Biotech
Seals Microplate Seals
Ideal for cold storage, moderate and high temperature incubation, protection of light sensitive samples and high throughput samples.
Strong 2.7 mil thick pierceable aluminum film. Non-cytotoxic and sterile. Temperature stable from -80°C to 110°C. Resistant to DMSO, chemicals, solvents and other common reagents. Two scored end tabs for easy positioning
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty