Would you like to log in again?
ThermalSeal 2™ Sealing Films, Excel Scientific
Supplier: Excel Scientific
TS2-100
490007-370EA 78.6 USD InStock
490007-370
ThermalSeal 2™ Sealing Films, Excel Scientific
Seals Microplate Seals
These improved films for classic thermal cycling applications consist of a 50 m polyester film with a 41 m layer of acrylic adhesive.
  • Length without end tabs: 125.1 mm
  • Adhesive: Acrylic, 41 m thick
  • Film: Polyester, 50 m thick
  • Non-pierceable Properties
  • Certified DNase-, RNase-, and nucleic-acid-free
The ThermalSeal 2 films offer better adhesion, reduced evaporation, longer end tabs for easier handling, and less tendency to tear during removal of the plate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty