Would you like to log in again?
Aluminum PCR Sealing Foil, GeneMate
Supplier: GeneMate
AF-GBNL-100
490001-214PK 221.45 USD InStock
490001-214
Aluminum PCR Sealing Foil, GeneMate
Seals Microplate Seals
Ideal for PCR applications, storage of sample material and active agents.
  • Adheres well to polypropylene, polystyrene, and polycarbonate without special equipment
  • Compatible with aqueous solutions and organic solvents
  • Minimal evaporation and cross contamination
  • Temperature resistant from −80 °C to 110 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty