Would you like to log in again?
MBP Perkin-Elmer®/Packard®-Style Tips & Tecan®-Style Tips, Thermo Scientific
Supplier: Thermo Scientific
904-013
490008-522CS 1679.21 USD InStock
490008-522
MBP Perkin-Elmer®/Packard®-Style Tips & Tecan®-Style Tips, Thermo Scientific
Pipette Tips
Also for Thermo Orion®. Filtered, conductive tip style, liquid sensing.
  • For use with: Tecan™ Freedom EVO™ LiHa Head, Tecan™ Genesis™
  • Non sterile, made from PP
Perkin-Elmer is a registered trademark of Perkin-Elmer Corporation. ART is a registered trademark of MBP and is protected by U.S. Patent # 5,156,811. EconoPak is a trademark of Molecular BioProducts. Tecan is a registered trademark of Tecan AG. Cavro is a registered trademark of Scientific Instruments, Inc. MultiPROBE and Packard are registered trademarks of Packard Instrument Company, Inc. Thermo Orion is a registered trademark of Thermo Orion Inc.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty