Would you like to log in again?
Axygen® Platinum Low Retention Filtered and Non-Filtered Tips, Corning
Supplier: Corning
Axygen®
TF-1000-L-R-S T-1000-C-L-R-S
490013-240CS 443.75 USD InStock
490013-240 490013-238
Axygen® Platinum Low Retention Filtered and Non-Filtered Tips, Corning
Pipette Tips
Platinum Low Retention plastics are universal fit, clear-metal free and designed for technical research.
  • Ultra-low retention, Inertno additives
  • Free of additives and are inert
  • Provides better quantitative results
  • Wall surfaces are free of occlusions and cavities which allows 99.9% of the sample to be dispensed
  • Platinum plastics are certified DNase-, RNase- and Pyrogen-free
Platinum Low Retention Plastics are made without costly, messy, and inconsistent processes such as adding silicone. The manufacturing process involves a change to the composition of the polymer which smoothes wall surfaces without chemical additives.
Filter Fits Pipetman, Eppendorf, Labsystems, Oxford, Biohit® , Sealpette, and GeneMate pipettors. Non-filtered Fits: Pipetman, Eppendorf, Labsystems, Oxford, Biohit® , Sealpette, GeneMate pipettors. Platinum, Yellow, and Blue pipette Tips Fit: Pipetman, Eppendorf, Labsystems, Oxford, Biohit® , Sealpette, GeneMate pipettors.
Eppendorf Research is a registered trademark of Eppendorf-Netheler-Hinz GmbH. Pipetman is a registered trademark of Rainin Instrument Co., Inc. Labsystems is a registered trademark of Labsystems Oy Corp. Biohit is a registered trademark of Biohit Oy Corp.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty