Would you like to log in again?
GeneMate Individual 0.2 ml PCR Tubes, Scientific Specialties
Supplier: Scientific Specialties
3110-00-110IS 3123-00-410IS
490003-560BX 120.05 USD InStock
490003-560 490003-592
GeneMate Individual 0.2 ml PCR Tubes, Scientific Specialties
Tubes PCR Tubes
Available packaged tube only, with 8- or 12-strip caps.
  • Capless tubes are the same premium quality as our PCR tubes with attached caps
  • Feature a smooth capless rim
Certified RNase-, DNase- and pyrogen-free.
Packaging: 1000 tubes/pack
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty