Would you like to log in again?
Laser Printer Labels, Laser Cryo-Tags® / Laser Cryo-Babies®, Diversified Biotech
Supplier: Diversified Biotech
LCRY-1200-R RNBW-2100 LCRY-1200-G LCRY-1200-B RNBW-2200 LCRY-1200-O LCRY-1200-Y
490006-812PK 149.25 USD InStock
490006-812 490006-922 490006-810 490006-814 490006-808 MSPP-RNDW-2200 490006-806
Laser Printer Labels, Laser Cryo-Tags® / Laser Cryo-Babies®, Diversified Biotech
Labels
Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers.
  • Chemically inert, polyolefin labels resist most organic solvents and caustic agents
  • Adhere to most plastics, glass, and metals without cracking, peeling, or degrading
  • Will fit a wide variety of cryovials, microtubes, storage boxes and other containers
  • Available in different colours
Heat resistant from -196 °C to 100 °C: Cryo-Babies and Cryo-Tags withstand boiling water baths, moderate oven temperatures (150°C), autoclaving and extreme freezer temperatures (liquid and vapor phase nitrogen).
Ordering information: Rainbow packs contain 4 sheets each of: Blue, Green, Orange, Red, Yellow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty