Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GeneMate Magnetic Hotplate Stirrer

Supplier: Benchmark Scientific
H4000-HS-GM
490000-188EA 1041.16 USD InStock
490000-188
GeneMate Magnetic Hotplate Stirrer
Stirrers Magnetic Hotplate Stirrers
Magnetic hotplate stirrer that is compatible with a wide variety of borosilicate glass beakers, flasks, bottles and other vessels.

  • Ceramic work surface, 7.5×7.5"
  • Stir speed from 60 to 1500 rpm
  • Temperature control up to 380 °C
  • Space efficient design (8×9") facilitates use on crowded benchtops and/or inside bio-hoods
  • Durable, chemical resistant white ceramic work surface

GeneMate magnetic hotplate-stirrers feature an exceptionally durable, chemical resistant white ceramic work surface. Their space-efficient design (8×9" footprint), makes them ideal for use on crowded bench tops and inside of bio-hoods.

Advanced microprocessor controls with convenient turn knobs allow quick, precise adjustment and maintenance of speed and temperature. Safety indicator LED's on the front panel indicate when the heating and/or stirring functions have been activated. With a square 7.5" in work surface, the units are compatible with a wide variety of popular sizes of borosilicate glass beakers, flasks, bottles, and other vessels.

Corresponding magnetic stirrers (stirring only 490000-192) or hotplates (heating only 490000-184) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.

Delivery information: Supplied complete with a support rod for mounting thermometers and temperature probes (490000-188 only)

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty