Would you like to log in again?
Contact Dish With Grid, Bioplast Manufacturing
Supplier: Bioplast Manufacturing
2911
490005-222CS 328.69 USD InStock
490005-222
Contact Dish With Grid, Bioplast Manufacturing
Culture Dishes Petri Dishes
Guaranteed Sterilized via Gamma Irradiation Process. Precision & Convenience. Designed to determine bio-burden counts on working surfaces. Grid consists of 16 (10x10 mm) squares marked numerically in one direction and alphabetically in the other. Media well permits dish and media to be inverted and placed in contact with working surface. Unique outer ring construction prevents contamination from fingers touching the media. Suitable for all automated filling equipment.
  • Stackable construction
  • Made for easy handling
  • Ventilation ribs minimize condensation
  • Domed base reduces media costs
  • Guaranteed sterile via gamma irradiation
  • Disposable
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty