Would you like to log in again?
Duran Screw Caps and Pouring Rings, Wheaton
Supplier: DWK Life Sciences
292422809 292392809 292391907 292421907
490009-888PK 15.92 USD InStock
490009-888 490009-884 490009-886 490009-890
Duran Screw Caps and Pouring Rings, Wheaton
Closures Bottle Closures
Blue polypropylene closures are linerless and fully autoclavable
  • Autoclavable
  • Chemically resistant
Caps are resistant to acids, alkalis, and commonly used solvents. Caps are for use with B-5005 series bottles. Blue and red pouring rings are easy to replace and are for use with the linerless and high-temperature screw caps listed below.
Red high-temp caps are teflon lined and are for high temperature applications
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty