Would you like to log in again?
GeneMate Full-Skirted, Low-Profile, 96-Well PCR Plates, Scientific Specialties
Supplier: Scientific Specialties
3441-00-520IS
490003-844BX 128.24 USD InStock
490003-844
GeneMate Full-Skirted, Low-Profile, 96-Well PCR Plates, Scientific Specialties
Microplates PCR Plates
Compatible with most commercially available thermal cyclers.
  • Full-skirt design reduces warping caused by thermal cycling
  • Ultra-thin wall for optimum heat transfer and higher yield
  • High contrast black alphanumerics for rapid sample identification
  • Plates can be sealed with strip caps, sealing mats, or sealing film
  • Crystal-clear clarity
  • Easy to visualize content of each well
  • Certified free from DNase, RNase, and detectable DNA
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty