Would you like to log in again?
GeneMate Sterile Centrifuge Tubes, Greiner bio-one
Supplier: Greiner bio-one
188261-ISC 188271-ISC
490001-621CS 357.27 USD InStock
490001-621 490001-623
GeneMate Sterile Centrifuge Tubes, Greiner bio-one
Tubes Centrifuge Tubes
These sterile tubes are free of detectable DNase/RNase/human DNA.
  • 15000 ×g Maximum RCF (15 ml Tubes)
  • 9500 ×g Maximum RCF (50 ml Tubes)
  • Polyproplene
  • Sterile
  • Racked or bulk packaging
  • Non-pyrogenic/Non-cytotoxic
Manufactured from high quality, crystal-clear polypropylene. Tubes feature white printed graduations and a shaded writing area. The 15 ml tube rack can be recycled.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty