Would you like to log in again?
SP Bel-Art Hot Hand® Protector, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
F38000-0000
490012-374CS 867.51 USD InStock
490012-374
SP Bel-Art Hot Hand® Protector, Bel-Art Products, a part of SP
Gloves
Safe, non-slip protection against very hot or cold surfaces.
  • Secure grip on hot or cold surfaces
  • Durable silicone rubber construction
  • Flexible over a range of extreme temperatures
Measures 3⁷/₈"× 7¹/₂" (10×19 cm) with durable silicone rubber construction that remains flexible from –57 °C to 260 °C (-70 °F to 500 °F). End pockets accept thumb and fingers to grasp objects. Multiple studs with concave tips cover surface of the mitt, providing secure grip on virtually all hot or cold surfaces.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty