Would you like to log in again?
E-Grip® Powder-Free Latex Examination Gloves, Microflex®
Supplier: Ansell
L971 L970 L972 L973 L974
10119-021BX 22.3 USD InStock
10119-021 10119-023 10119-014 10119-017 10119-019
E-Grip® Powder-Free Latex Examination Gloves, Microflex®
Gloves
The micro-textured satin finish of E-Grip gloves provides enhanced gripping power and excellent tactile sensitivity.
  • Beaded cuffs for easy donning
  • 510K approved for medical use
Gloves are nonsterile and they feature textured fingers for improved tactile sensitivity and strength.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty